Gene Transcripts

Gene Symbol
4933417A18Rik
Entrez Gene ID
66761
Gene Name
RIKEN cDNA 4933417A18 gene
Gene Aliases
Chromosome Location
Chr. 13: 34924405 - 34956218 on Build GRCm38
UniGene ID
Mm.157047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025750.3 NP_080026.2 2, 3 510
GenBank mRNA AK016838.1 - 2, 3 511
BC100450.1 - 2, 3 500 30919421
BC145747.1 - 2, 3 465 40131050

View Full Product Details