Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000239.2 | NP_000230.1 | 1 | 131 | |
GenBank mRNA | AK130149.1 | - | 1 | 136 | |
AK310930.1 | - | 113 | |||
AK311779.1 | - | 1 | 104 | ||
BC004147.2 | - | 1 | 95 | 2959387 | |
FJ795023.1 | - | 1 | 98 | ||
J03801.1 | - | 1 | 101 | ||
KJ841928.1 | - | 1 | 76 | ||
M19045.1 | - | 1 | 89 | ||
M21119.1 | - | 1 | 89 | ||
U25677.1 | - | 1 | 76 |