Gene Transcripts
- Gene Symbol
- Pnlip
- Entrez Gene ID
- 69060
- Gene Name
- pancreatic lipase
- Gene Aliases
- 1810007A24Rik, PL, PTL
- Chromosome Location
- Chr. 19: 58670365 - 58681788 on Build GRCm38
- UniGene ID
- Mm.20407
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007357.1 |
- |
5 |
496 |
|
View Full Product Details