Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163579.1 | NP_001157051.1 | 5 | 803 | |
NM_001163580.1 | NP_001157052.1 | 5 | 803 | ||
NM_028915.3 | NP_083191.2 | 5 | 803 | ||
GenBank mRNA | AK004646.1 | - | 5 | 804 | |
AK016559.1 | - | 5 | 742 | ||
AK029563.1 | - | 5 | 720 | ||
AK173256.1 | - | 5 | 731 | ||
BC061202.1 | - | 5 | 759 | ||
BC099680.1 | - | 5 | 767 | ||
BC138734.1 | - | 5 | 777 | 8861756 | |
BC138735.1 | - | 5 | 777 | 8861757 |