Gene Transcripts
- Gene Symbol
- Pabpc4
- Entrez Gene ID
- 230721
- Gene Name
- poly(A) binding protein, cytoplasmic 4
- Gene Aliases
- Chromosome Location
- Chr. 4: 123262387 - 123298832 on Build GRCm38
- UniGene ID
- Mm.277091
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC003283.1 |
- |
7 |
1191 |
|
View Full Product Details