Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134437.3 | NP_602319.1 | 3 | 384 | |
GenBank mRNA | AF459444.1 | - | 3 | 303 | |
AF494210.1 | - | 3 | 444 | ||
AK133233.1 | - | 5 | 547 | ||
BC066804.1 | - | 3 | 306 | ||
BC138629.1 | - | 3 | 339 | 8861649 |