Gene Transcripts

Gene Symbol
PRLR
Entrez Gene ID
5618
Gene Name
prolactin receptor
Gene Aliases
HPRL, MFAB, hPRLrI
Chromosome Location
Chr. 5: 35048295 - 35230589 on Build GRCh38
UniGene ID
Hs.368587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000949.6 NP_000940.1 4 558
NM_001204315.1 NP_001191244.1 3 236
NM_001204316.1 NP_001191245.1 3 236
NM_001204317.1 NP_001191246.1 3 236
NM_001204318.1 NP_001191247.1 3 236
XM_006714484.2 XP_006714547.1 4 376
XM_011514068.2 XP_011512370.1 4 391
XM_011514069.2 XP_011512371.1 4 330
XM_017009646.1 XP_016865135.1 2 174
GenBank mRNA AF166329.1 - 2 131
AF416618.1 - 2 148
AF416619.1 - 2 148
AF492470.1 - 2 148
AK313270.1 - 4 446
BC059392.1 - 4 444 30343995
DA640476.1 - 4 391
GU133399.1 - 2 148
KJ651922.1 - 5 560
KJ651923.1 - 5 508
KJ651924.1 - 4 417
KJ651925.1 - 4 424
M31661.1 - 4 415
S78505.1 - 1 59

View Full Product Details