Gene Transcripts

Gene Symbol
Tatdn1
Entrez Gene ID
69694
Gene Name
TatD DNase domain containing 1
Gene Aliases
2310079P03Rik, CDA11
Chromosome Location
Chr. 15: 58890153 - 58933781 on Build GRCm38
UniGene ID
Mm.383230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175151.4 NP_780360.1 12 879
GenBank mRNA AK042248.1 - 12 880

View Full Product Details