Gene Transcripts

Gene Symbol
Adamts9
Entrez Gene ID
101401
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 9
Gene Aliases
1810011L16Rik, 8430403M15Rik, AW743315, E030027K14Rik
Chromosome Location
Chr. 6: 92772699 - 92944047 on Build GRCm38
UniGene ID
Mm.158235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175314.3 NP_780523.2 22 3291
GenBank mRNA AK030699.1 - 9 1401
AK078802.1 - 4 486
BC068142.1 - 8 1330

View Full Product Details