Gene Transcripts
- Gene Symbol
- Cit
- Entrez Gene ID
- 12704
- Gene Name
- citron
- Gene Aliases
- C030025P15Rik, CRIK, CRIK-SK, Cit-k
- Chromosome Location
- Chr. 5: 115845224 - 116008586 on Build GRCm38
- UniGene ID
- Mm.426282
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK047894.1 |
- |
1 |
982 |
|
View Full Product Details