Gene Transcripts

Gene Symbol
Dnah2
Entrez Gene ID
327954
Gene Name
dynein, axonemal, heavy chain 2
Gene Aliases
2900022L05Rik, 4930564A01, D130094J20, D330014H01Rik, Dnahc2, Dnhd3
Chromosome Location
Chr. 11: 69420807 - 69549108 on Build GRCm38
UniGene ID
Mm.247478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081330.1 NP_001074799.1 59, 60 9427
GenBank mRNA AK029791.1 - 6, 7 900

View Full Product Details