Gene Transcripts

Gene Symbol
Igsf1
Entrez Gene ID
209268
Gene Name
immunoglobulin superfamily, member 1
Gene Aliases
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364
Chromosome Location
Chr. X: 49782536 - 49797906 on Build GRCm38
UniGene ID
Mm.110505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177915.4 NP_808583.1 5 1358
GenBank mRNA AK030452.1 - 5 1330
AY227772.1 - 5 1352
BC089474.1 - 5 1339
BC100420.1 - 5 1358

View Full Product Details