Gene Transcripts

Gene Symbol
EN2
Entrez Gene ID
2020
Gene Name
engrailed homeobox 2
Gene Aliases
Chromosome Location
Chr. 7: 155458129 - 155464831 on Build GRCh38
UniGene ID
Hs.134989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001427.3 NP_001418.2 2 1155
GenBank mRNA BC104970.1 - 2 1155
BC104972.1 - 2 1155 8143975

View Full Product Details