Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002231.2 | NP_001002231.1 | 4 | 619 | |
NM_001256080.1 | NP_001243009.1 | 3 | 459 | ||
NM_005551.4 | NP_005542.1 | 4 | 619 | ||
NR_045762.1 | 4 | 606 | |||
NR_045763.1 | 4 | 705 | |||
GenBank mRNA | AF188745.1 | - | 4 | 581 | |
AF188746.1 | - | 4 | 594 | ||
AF188747.1 | - | 4 | 594 | ||
AK300527.1 | - | 4 | 705 | ||
AK300549.1 | - | 3 | 453 | ||
AK300566.1 | - | 4 | 613 | ||
AK309968.1 | - | 2 | 581 | ||
AK309969.1 | - | 4 | 545 | ||
AK309973.1 | - | 3 | 440 | ||
AK309982.1 | - | 2 | 572 | ||
AK309985.1 | - | 3 | 456 | ||
AK315890.1 | - | 3 | 433 | ||
AY429509.1 | - | 3 | 720 | ||
AY429510.1 | - | 2 | 560 | ||
BC005196.1 | - | 4 | 619 | 3951036 | |
BT006650.1 | - | 4 | 578 | ||
S39329.1 | - | 4 | 594 |