Gene Transcripts
- Gene Symbol
- LYZ
- Entrez Gene ID
- 4069
- Gene Name
- lysozyme
- Gene Aliases
- LYZF1, LZM
- Chromosome Location
- Chr. 12: 69348354 - 69354233 on Build GRCh38
- UniGene ID
- Hs.524579
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000239.2 |
NP_000230.1
|
2, 3 |
358 |
|
GenBank mRNA |
AK130149.1 |
- |
2, 3 |
363 |
|
AK311779.1 |
- |
2, 3 |
331 |
|
BC004147.2 |
- |
2, 3 |
322 |
2959387 |
FJ795023.1 |
- |
2, 3 |
325 |
|
J03801.1 |
- |
2, 3 |
328 |
|
KJ841928.1 |
- |
2, 3 |
303 |
|
M19045.1 |
- |
2, 3 |
316 |
|
M21119.1 |
- |
2, 3 |
316 |
|
U25677.1 |
- |
2, 3 |
303 |
|
View Full Product Details