Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304722.1 | NP_001291651.1 | 2 | 482 | |
NM_001304723.1 | NP_001291652.1 | 1 | 455 | ||
NM_001304724.1 | NP_001291653.1 | 2 | 482 | ||
NM_016274.5 | NP_057358.2 | 2 | 515 | ||
XM_017001420.1 | XP_016856909.1 | 2 | 440 | ||
GenBank mRNA | AF073836.1 | - | 1 | 145 | |
AF168676.1 | - | 2 | 247 | ||
AF192912.1 | - | 2 | 452 | ||
AF217956.1 | - | 2 | 303 | ||
AF291105.1 | - | 2 | 443 | ||
AK222921.1 | - | 2 | 159 | ||
AK302255.1 | - | 2 | 499 | ||
BC010149.2 | - | 2 | 243 | 4330014 | |
BC023533.2 | - | 2 | 339 | ||
BP329810.1 | - | 1 | 204 | ||
BX353413.2 | - | 2 | 483 | ||
BX376480.2 | - | 2 | 451 | ||
BX394578.2 | - | 2 | 381 | ||
DA604331.1 | - | 2 | 351 | ||
DB023901.1 | - | 1 | 455 |