Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017564.9 | NP_060034.9 | 12 | 1514 | |
XM_011538537.2 | XP_011536839.1 | 12 | 1555 | ||
XM_011538538.2 | XP_011536840.1 | 12 | 1555 | ||
XM_017019585.1 | XP_016875074.1 | 12 | 1555 | ||
GenBank mRNA | AB052958.1 | - | 12 | 1510 | |
AJ295695.1 | - | 12 | 1514 | ||
AK160380.1 | - | 11 | 1229 | ||
AY311388.1 | - | 12 | 1532 |