Gene Transcripts
- Gene Symbol
- STK40
- Entrez Gene ID
- 83931
- Gene Name
- serine/threonine kinase 40
- Gene Aliases
- SHIK, SgK495
- Chromosome Location
- Chr. 1: 36339619 - 36385927 on Build GRCh38
- UniGene ID
- Hs.471768
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282546.1 |
NP_001269475.1
|
6, 7 |
850 |
|
NM_001282547.1 |
NP_001269476.1
|
6, 7 |
835 |
|
NM_032017.2 |
NP_114406.1
|
7, 8 |
1072 |
|
GenBank mRNA |
AK024504.1 |
- |
5, 6 |
634 |
|
AK075029.1 |
- |
3, 4 |
351 |
|
AK075048.1 |
- |
6, 7 |
829 |
|
AL834137.1 |
- |
7, 8 |
1029 |
|
AY198395.1 |
- |
6, 7 |
805 |
|
BC005169.2 |
- |
1, 2 |
34 |
|
BC007835.2 |
- |
6, 7 |
803 |
4109652 |
BC008344.1 |
- |
6, 7 |
811 |
3530483 |
View Full Product Details