Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004419.4 | NP_001004419.1 | 3 | 257 | |
NM_001197317.2 | NP_001184246.1 | 2 | 146 | ||
NM_001197318.2 | NP_001184247.1 | 3 | 257 | ||
NM_001197319.2 | NP_001184248.1 | 2 | 146 | ||
NM_013269.5 | NP_037401.1 | 3 | 257 | ||
NR_036693.2 | 3 | 257 | |||
GenBank mRNA | AF133299.1 | - | 3 | 243 | |
AF285087.1 | - | 3 | 235 | ||
AF285088.1 | - | 2 | 152 | ||
AF285089.1 | - | 2 | 167 | ||
AK310150.1 | - | 3 | 253 | ||
AK310554.1 | - | 3 | 248 | ||
AK313554.1 | - | 3 | 248 | ||
BC019883.1 | - | 3 | 237 | 4524891 | |
BU929933.1 | - | 3 | 242 | ||
CB958277.1 | - | 3 | 254 | ||
DA814760.1 | - | 3 | 252 | ||
DB111977.1 | - | 3 | 258 | ||
DB124897.1 | - | 3 | 253 | ||
FN813349.1 | - | 3 | 230 | ||
FN813350.1 | - | 2 | 119 | ||
FN813351.1 | - | 2 | 119 | ||
KF958454.1 | - | 3 | 230 | ||
KF958455.1 | - | 3 | 230 | ||
KF958456.1 | - | 3 | 230 | ||
KF958457.1 | - | 3 | 230 | ||
KF958458.1 | - | 3 | 230 | ||
KF958459.1 | - | 2 | 119 | ||
KF971856.1 | - | 3 | 230 | ||
KF971857.1 | - | 3 | 230 | ||
KF971858.1 | - | 2 | 119 | ||
KF971859.1 | - | 3 | 230 |