Gene Transcripts
- Gene Symbol
- FLJ20021
- Entrez Gene ID
- 90024
- Gene Name
- uncharacterized LOC90024
- Gene Aliases
- Chromosome Location
- Chr. 4: 101347777 - 101348883 on Build GRCh38
- UniGene ID
- Hs.534513
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033874.1 |
|
2 |
287 |
|
GenBank mRNA |
AK000028.1 |
- |
2 |
284 |
|
View Full Product Details