Gene Transcripts

Gene Symbol
AP3M1
Entrez Gene ID
26985
Gene Name
adaptor related protein complex 3 mu 1 subunit
Gene Aliases
Chromosome Location
Chr. 10: 74120257 - 74151085 on Build GRCh38
UniGene ID
Hs.500104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320263.1 NP_001307192.1 4 638
NM_001320264.1 NP_001307193.1 2 477
NM_001320265.1 NP_001307194.1 2 351
NM_012095.5 NP_036227.1 2 351
NM_207012.3 NP_996895.1 3 477
NR_135191.1 2 351
GenBank mRNA AF092092.1 - 1 17
AK026983.1 - 2 86
AK299352.1 - 2 102
AK315597.1 - 2 60
BC026232.1 - 3 236 4807259
BC067127.1 - 2 447 6169822
BP230128.1 - 2 336
BU172918.1 - 2 315
BU430175.1 - 2 336
DC323908.1 - 4 376
DC418192.1 - 2 304

View Full Product Details