Gene Transcripts
- Gene Symbol
- CNTN6
- Entrez Gene ID
- 27255
- Gene Name
- contactin 6
- Gene Aliases
- NB3
- Chromosome Location
- Chr. 3: 978021 - 1403610 on Build GRCh38
- UniGene ID
- Hs.387300
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289080.1 |
NP_001276009.1
|
4, 5 |
894 |
|
NM_001289081.1 |
NP_001276010.1
|
3, 4 |
757 |
|
NM_014461.3 |
NP_055276.1
|
4, 5 |
903 |
|
XM_005265058.3 |
XP_005265115.1
|
5, 6 |
1026 |
|
XM_011533590.2 |
XP_011531892.1
|
4, 5 |
550 |
|
XM_011533591.2 |
XP_011531893.1
|
3, 4 |
865 |
|
XM_017006171.1 |
XP_016861660.1
|
9, 10 |
1453 |
|
XM_017006172.1 |
XP_016861661.1
|
4, 5 |
1028 |
|
XM_017006173.1 |
XP_016861662.1
|
4, 5 |
642 |
|
XM_017006174.1 |
XP_016861663.1
|
3, 4 |
517 |
|
XM_017006175.1 |
XP_016861664.1
|
2, 3 |
294 |
|
XM_017006176.1 |
XP_016861665.1
|
4, 5 |
642 |
|
XM_017006177.1 |
XP_016861666.1
|
4, 5 |
642 |
|
GenBank mRNA |
AB003592.1 |
- |
4, 5 |
616 |
|
AK294786.1 |
- |
3, 4 |
757 |
|
AK307814.1 |
- |
5, 6 |
643 |
|
AK314220.1 |
- |
4, 5 |
617 |
|
BC113118.1 |
- |
4, 5 |
529 |
40036214 |
View Full Product Details