Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002231.2 | NP_001002231.1 | 4 | 645 | |
NM_001256080.1 | NP_001243009.1 | 3 | 485 | ||
NM_005551.4 | NP_005542.1 | 4 | 645 | ||
NR_045762.1 | 4 | 632 | |||
NR_045763.1 | 4 | 731 | |||
GenBank mRNA | AF188745.1 | - | 4 | 607 | |
AF188746.1 | - | 4 | 620 | ||
AF188747.1 | - | 4 | 620 | ||
AK300527.1 | - | 4 | 731 | ||
AK300549.1 | - | 3 | 479 | ||
AK300566.1 | - | 4 | 639 | ||
AK309968.1 | - | 2 | 607 | ||
AK309969.1 | - | 4 | 571 | ||
AK309973.1 | - | 3 | 466 | ||
AK309982.1 | - | 2 | 598 | ||
AK309985.1 | - | 3 | 482 | ||
AK315890.1 | - | 3 | 459 | ||
AY429509.1 | - | 3 | 746 | ||
AY429510.1 | - | 2 | 586 | ||
BC005196.1 | - | 4 | 645 | 3951036 | |
BT006650.1 | - | 4 | 604 | ||
S39329.1 | - | 4 | 620 |