Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014080.4 | NP_054799.4 | 29 | 3937 | |
XM_005254421.2 | XP_005254478.1 | 29 | 3937 | ||
GenBank mRNA | AF181972.1 | - | 21 | 2720 | |
AF230496.1 | - | 29 | 3919 | ||
AF267981.1 | - | 29 | 3937 |