Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 23161883-23161339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA8CP
Entrez Gene ID
729786
Gene Name
golgin A8 family member C, pseudogene
Gene Aliases
GOLGA8C
UniGene
Hs.592025
Gene Chromosome Location
Chr.15: 20562349-20575703 on Build GRCh38
Transcripts
NR_027411.2 NR_027407.1

Gene Symbol
GOLGA8DP
Entrez Gene ID
100132979
Gene Name
golgin A8 family member D, pseudogene
Gene Aliases
GOLGA8D
UniGene
Hs.454647
Gene Chromosome Location
Chr.15: 23157340-23170783 on Build GRCh38
Transcripts
NR_027411.2 NR_027407.1

View Full Product Details