Gene Transcripts

Gene Symbol
Arhgef6
Entrez Gene ID
73341
Gene Name
Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6
Gene Aliases
1600028C08Rik, 1700038J06Rik, 4930592P22Rik, alpha-PIX
Chromosome Location
Chr. X: 57231485 - 57338756 on Build GRCm38
UniGene ID
Mm.261443
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152801.2 NP_690014.2 5 845
GenBank mRNA AF393831.1 - 5 845
AK016387.1 - 2 269
AK028642.1 - 4 860
AK040468.1 - 5 667

View Full Product Details