Gene Transcripts

Gene Symbol
Vmn1r6
Entrez Gene ID
171193
Gene Name
vomeronasal 1 receptor 6
Gene Aliases
V1rc20
Chromosome Location
Chr. 6: 57002355 - 57003266 on Build GRCm38
UniGene ID
Mm.377173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134175.1 NP_598936.1 1 796
GenBank mRNA BC125532.1 - 1 851
BC125534.1 - 1 851 40130049

View Full Product Details