Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166249.1 | NP_001159721.1 | 3 | 345 | |
NM_001166250.1 | NP_001159722.1 | 3 | 660 | ||
NM_001166251.1 | NP_001159723.1 | 3 | 660 | ||
NM_011844.4 | NP_035974.1 | 3 | 345 | ||
GenBank mRNA | AJ001118.2 | - | 3 | 273 | |
AJ316580.1 | - | 3 | 536 | ||
AK006949.1 | - | 3 | 656 | ||
AK028008.1 | - | 3 | 288 | ||
AK131645.1 | - | 3 | 325 | ||
AK148242.1 | - | 3 | 288 | ||
AK148395.1 | - | 3 | 288 | ||
AK185666.1 | - | 3 | 210 | ||
AK188627.1 | - | 3 | 210 | ||
AK190239.1 | - | 2 | 188 | ||
AK191795.1 | - | 2 | 188 | ||
AK196136.1 | - | 2 | 188 | ||
AK196912.1 | - | 2 | 188 | ||
AK197275.1 | - | 2 | 188 | ||
AK198198.1 | - | 2 | 188 | ||
AK198283.1 | - | 2 | 188 | ||
AK198733.1 | - | 2 | 188 | ||
AK199935.1 | - | 3 | 234 | ||
AK201810.1 | - | 2 | 189 | ||
AK202498.1 | - | 2 | 187 | ||
AK203723.1 | - | 2 | 185 | ||
AK206822.1 | - | 3 | 210 | ||
AK208240.1 | - | 3 | 210 | ||
AK211363.1 | - | 3 | 210 | ||
AK215869.1 | - | 3 | 255 | ||
BC057965.1 | - | 3 | 270 | 4986023 | |
BY706980.1 | - | 3 | 664 |