Gene Transcripts

Gene Symbol
Aldob
Entrez Gene ID
230163
Gene Name
aldolase B, fructose-bisphosphate
Gene Aliases
Aldo-2, Aldo2, BC016435
Chromosome Location
Chr. 4: 49535995 - 49549483 on Build GRCm38
UniGene ID
Mm.482116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144903.2 NP_659152.1 4 413
GenBank mRNA AK135385.1 - 4 442
AK149395.1 - 4 430
AK149598.1 - 4 427
AK167334.1 - 4 430
AK167566.1 - 4 427
AK168600.1 - 4 427
AK168997.1 - 4 477
BC016435.1 - 4 409 4241025
BC022113.1 - 4 386 5098524
BC024056.1 - 4 419 5100067
BC024112.1 - 4 418 5102118
BC026577.1 - 4 378 4989521
BC030724.1 - 4 415 5100784
BC030725.1 - 4 419 5101686
BC034169.1 - 4 419 4975382
BC034171.1 - 4 396 4972521
BC034172.1 - 4 383 4974489
BC034173.1 - 4 395 4974553
BC036130.1 - 4 396 4974071
BC036131.1 - 4 413 4974877
BC036132.1 - 4 413 4975336
BC036133.1 - 4 425 4975573

View Full Product Details