Gene Transcripts
- Gene Symbol
- Gm26777
- Entrez Gene ID
- 102632019
- Gene Name
- predicted gene, 26777
- Gene Aliases
- Chromosome Location
- Chr. 12: 80509945 - 80518157 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK045378.1 |
- |
|
1301 |
|
- Gene Symbol
- 2310002D06Rik
- Entrez Gene ID
- 69522
- Gene Name
- RIKEN cDNA 2310002D06 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 80507206 - 80517954 on Build GRCm38
- UniGene ID
- Mm.390530
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009088.1 |
- |
3 |
168 |
|
View Full Product Details