Gene Transcripts

Gene Symbol
1700030F18Rik
Entrez Gene ID
72277
Gene Name
RIKEN cDNA 1700030F18 gene
Gene Aliases
FAM186A
Chromosome Location
Chr. 15: 99918345 - 99967430 on Build GRCm38
UniGene ID
Mm.272766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028180.3 NP_082456.1 2 262
GenBank mRNA AK006542.1 - 2 261
BC048613.1 - 2 262 6742336

View Full Product Details