Gene Transcripts

Gene Symbol
4930550L24Rik
Entrez Gene ID
75352
Gene Name
RIKEN cDNA 4930550L24 gene
Gene Aliases
mage-k1
Chromosome Location
Chr. X: 58911461 - 58920304 on Build GRCm38
UniGene ID
Mm.60906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023774.3 NP_076263.1 2 561
GenBank mRNA AF319984.1 - 1 411
AK016090.1 - 2 539
BC087920.1 - 2 561 30281980

View Full Product Details