Gene Transcripts

Gene Symbol
Pi4k2a
Entrez Gene ID
114554
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Gene Aliases
Pi4KII
Chromosome Location
Chr. 1: 261337594 - 261362509 on Build Rnor_6.0
UniGene ID
Rn.162533
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053735.1 NP_446187.1 7 1171
GenBank mRNA AY029199.1 - 7 1171

View Full Product Details