Gene Transcripts

Gene Symbol
Fam175a
Entrez Gene ID
289468
Gene Name
family with sequence similarity 175, member A
Gene Aliases
Ccdc98, RGD1305287
Chromosome Location
Chr. 14: 10424674 - 10439010 on Build Rnor_6.0
UniGene ID
Rn.106875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009633.1 NP_001009633.1 7 675
GenBank mRNA BC088408.1 - 7 675

View Full Product Details