Gene Transcripts

Gene Symbol
Dhcr24
Entrez Gene ID
298298
Gene Name
24-dehydrocholesterol reductase
Gene Aliases
Chromosome Location
Chr. 5: 126164708 - 126188926 on Build Rnor_6.0
UniGene ID
Rn.225146
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080148.1 NP_001073617.1 8, 9 1398
GenBank mRNA AY310319.1 - 8, 9 1398
AY921220.2 - 8, 9 1460

View Full Product Details