Gene Transcripts

Gene Symbol
Rgsl2h
Entrez Gene ID
364021
Gene Name
regulator of G-protein signaling like 2 homolog (mouse)
Gene Aliases
Chromosome Location
Chr. 13: 71281410 - 71282417 on Build Rnor_6.0
UniGene ID
Rn.107065
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198790.1 NP_942085.1 1 336
GenBank mRNA AJ515383.1 - 1 336

View Full Product Details