Gene Transcripts

Gene Symbol
Ywhaq
Entrez Gene ID
25577
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta
Gene Aliases
14-3-3t
Chromosome Location
Chr. 6: 43463294 - 43493816 on Build Rnor_6.0
UniGene ID
Rn.2502
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013053.1 NP_037185.1 2 97
GenBank mRNA BC062409.1 - 1 67
D17614.1 - 2 97

View Full Product Details