Gene Transcripts

Gene Symbol
Copz1
Entrez Gene ID
315345
Gene Name
coatomer protein complex, subunit zeta 1
Gene Aliases
Chromosome Location
Chr. 7: 144900091 - 144926299 on Build Rnor_6.0
UniGene ID
Rn.159796
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108117.1 NP_001101587.1 3, 4 220
GenBank mRNA FQ226451.1 - 3, 4 196
FQ234671.1 - 3, 4 196

View Full Product Details