Gene Transcripts

Gene Symbol
Fas
Entrez Gene ID
246097
Gene Name
Fas cell surface death receptor
Gene Aliases
Tnfrsf6
Chromosome Location
Chr. 1: 252589785 - 252624790 on Build Rnor_6.0
UniGene ID
Rn.162521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139194.2 NP_631933.2 7, 8 676
GenBank mRNA D26113.1 - 7, 8 701
FQ233018.1 - 7, 8 664

View Full Product Details