Gene Transcripts

Gene Symbol
Olr1234
Entrez Gene ID
300558
Gene Name
olfactory receptor 1234
Gene Aliases
ratchr8-40553209-40554150_ORF
Chromosome Location
Chr. 4: 1459363 - 1460304 on Build Rnor_6.0
UniGene ID
Rn.143535
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000446.1 NP_001000446.1 1 457

View Full Product Details