Gene Transcripts

Gene Symbol
Edn3
Entrez Gene ID
366270
Gene Name
endothelin 3
Gene Aliases
Et3, RGD1564825, preproET-3
Chromosome Location
Chr. 3: 172856730 - 172879276 on Build Rnor_6.0
UniGene ID
Rn.201692
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077650.1 NP_001071118.1 2, 3 406
GenBank mRNA S39779.1 - 2, 3 406

View Full Product Details