Gene Transcripts

Gene Symbol
Acta2
Entrez Gene ID
81633
Gene Name
actin, alpha 2, smooth muscle, aorta
Gene Aliases
Chromosome Location
Chr. 1: 252537614 - 252550394 on Build Rnor_6.0
UniGene ID
Rn.195319
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031004.2 NP_112266.1 4 364
GenBank mRNA BC158550.1 - 4 358
DN934985.1 - 4 393
X06801.1 - 4 349

View Full Product Details