Gene Transcripts

Gene Symbol
GOLGA8F
Entrez Gene ID
100132565
Gene Name
golgin A8 family member F
Gene Aliases
-
Chromosome Location
Chr.15: 28378638 - 28391266 on Build GRCh38
Uni Gene
Hs.510812
Interrogated Sequence Translated Protein
RefSeq NR_033351.1
GenBank mRNA AK057241.1 -
AK302625.1 -
AL833871.1 -
BC144672.1 -
BC150302.1 -
DB071191.1 -
DQ309038.1 -
ENST00000532622.0 -

View Full Product Details