Gene Transcripts
- Gene Symbol
- Nkx6-2
- Entrez Gene ID
- 14912
- Gene Name
- NK6 homeobox 2
- Gene Aliases
- Gtx, Nkx6.2
- Chromosome Location
- Chr. 7: 139579376 - 139582797 on Build GRCm38
- UniGene ID
- Mm.28308
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027857.1 |
|
3 |
1381 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF471361.1 |
- |
1 |
241 |
|
Chr. 7 - 139581131 - 139581385 [-] |
View Full Product Details