Gene Transcripts

Gene Symbol
SOX2-OT
Entrez Gene ID
347689
Gene Name
SOX2 overlapping transcript
Gene Aliases
NCRNA00043, SOX2OT
Chromosome Location
Chr. 3: 181056680 - 181742228 on Build GRCh38
UniGene ID
Hs.654932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_004053.3 5 2345
NR_075089.1 4 2237
NR_075090.1 3 1951
NR_075091.1 8 2360
NR_075092.1 7 2252
NR_075093.1 5 1942
GenBank mRNA AK022826.1 - 5 2132
AL157425.1 - 3 1842
JN711430.1 - 4 2208
JN882275.1 - 3 1922
JQ408703.1 - 8 2359
JX110620.1 - 7 2451
JX504683.1 - 8 2413
KF981435.1 - 4 2059

View Full Product Details