Gene Transcripts

Gene Symbol
LINC00575
Entrez Gene ID
439934
Gene Name
long intergenic non-protein coding RNA 575
Gene Aliases
C4orf11
Chromosome Location
Chr. 4: 82613113 - 82621437 on Build GRCh38
UniGene ID
Hs.457189
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024087.1 3 501
NR_024088.1 3 439
GenBank mRNA AY316301.1 - 3 501
AY316302.1 - 3 439

View Full Product Details