Gene Transcripts
- Gene Symbol
- CRX
- Entrez Gene ID
- 1406
- Gene Name
- cone-rod homeobox
- Gene Aliases
- CORD2, CRD, LCA7, OTX3
- Chromosome Location
- Chr. 19: 47821842 - 47843330 on Build GRCh38
- UniGene ID
- Hs.617342
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF335593.1 |
- |
1 |
195 |
|
View Full Product Details