Gene Transcripts
- Gene Symbol
- KLK2
- Entrez Gene ID
- 3817
- Gene Name
- kallikrein related peptidase 2
- Gene Aliases
- KLK2A2, hGK-1, hK2
- Chromosome Location
- Chr. 19: 50873433 - 50880567 on Build GRCh38
- UniGene ID
- Hs.515560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF336106.1 |
- |
1 |
849 |
|
View Full Product Details