Gene Transcripts
- Gene Symbol
- Sox5os1
- Entrez Gene ID
- 71080
- Gene Name
- SRY (sex determining region Y)-box 5, opposite strand 1
- Gene Aliases
- 4933415D12Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.472043
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016816.1 |
- |
1 |
626 |
|
View Full Product Details