Gene Transcripts
- Gene Symbol
- 1700023F02Rik
- Entrez Gene ID
- 69433
- Gene Name
- RIKEN cDNA 1700023F02 gene
- Gene Aliases
- AV205046
- Chromosome Location
- Chr. 10: 66120609 - 66124064 on Build GRCm38
- UniGene ID
- Mm.23366
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038039.1 |
|
3 |
310 |
|
GenBank mRNA |
AK006272.1 |
- |
3 |
311 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706353.1 |
- |
3 |
311 |
|
Chr. 10 - 66120611 - 66124064 [-] |
View Full Product Details